The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244959.1 | NP_001231888.1 | 7 - 8 | 883 | 102 |
NM_001244960.1 | NP_001231889.1 | 7 - 8 | 659 | 102 | |
NM_001244961.1 | NP_001231890.1 | 2 - 3 | 195 | 102 | |
NM_174938.5 | NP_777598.3 | 7 - 8 | 883 | 102 | |
XM_017014588.1 | XP_016870077.1 | 7 - 8 | 573 | 102 | |
XM_017014589.1 | XP_016870078.1 | 2 - 3 | 3010 | 102 | |
XM_017014590.1 | XP_016870079.1 | 6 - 7 | 796 | 102 | |
GenBank mRNA | AK094281.1 | - | 7 - 8 | 584 | 102 |
AK223597.1 | - | 7 - 8 | 810 | 102 | |
AK297722.1 | - | 7 - 8 | 801 | 102 | |
AY137774.1 | - | 7 - 8 | 667 | 102 | |
BC037253.1 | - | 2 - 3 | 195 | 102 | |
EF560742.1 | - | 7 - 8 | 734 | 102 |