Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C12orf57
Entrez Gene ID
113246
Gene Name
chromosome 12 open reading frame 57
Gene Aliases
C10, GRCC10
Chromosome Location
Chr.12: 6943438 - 6946003 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301834.1 NP_001288763.1 2 - 3 226 65
NM_001301837.1 NP_001288766.1 1 - 2 355 65
NM_138425.3 NP_612434.1 1 - 2 355 65
GenBank mRNA AK311912.1 - 1 - 2 153 65
BC009925.2 - 1 - 2 132 65
BM687523.1 - 1 - 2 342 65
BM781587.1 - 1 - 2 355 65
CB161655.1 - 2 - 3 226 65

View Full Product Details