The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013371.3 | NP_037503.2 | 6 - 7 | 1381 | 75 |
NM_153758.2 | NP_715639.1 | 5 - 6 | 581 | 75 | |
GenBank mRNA | AF192498.1 | - | 5 - 6 | 489 | 75 |
AF453946.1 | - | 6 - 7 | 1381 | 75 | |
AY040367.1 | - | 5 - 6 | 581 | 75 | |
CA306526.1 | - | 1 - 2 | 75 |