The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040031.1 | NP_001035120.1 | 3 - 4 | 357 | 97 |
NM_001774.2 | NP_001765.1 | 3 - 4 | 391 | 97 | |
XM_005259435.3 | XP_005259492.1 | 3 - 4 | 393 | 97 | |
XM_005259436.3 | XP_005259493.1 | 3 - 4 | 389 | 97 | |
XM_011527542.2 | XP_011525844.1 | 3 - 4 | 354 | 97 | |
XM_011527543.2 | XP_011525845.1 | 3 - 4 | 459 | 97 | |
XM_011527544.2 | XP_011525846.1 | 3 - 4 | 428 | 97 | |
XM_017027513.1 | XP_016883002.1 | 3 - 4 | 389 | 97 | |
GenBank mRNA | AK058093.1 | - | 4 - 5 | 439 | 97 |
AK301016.1 | - | 3 - 4 | 381 | 97 | |
AK301328.1 | - | 3 - 4 | 384 | 97 | |
AK316348.1 | - | 3 - 4 | 281 | 97 | |
AL833693.1 | - | 2 - 3 | 1435 | 97 | |
BC106752.1 | - | 3 - 4 | 308 | 97 | |
BC110379.1 | - | 3 - 4 | 343 | 97 | |
BT019851.1 | - | 3 - 4 | 270 | 97 | |
CR541877.1 | - | 3 - 4 | 270 | 97 | |
X14046.1 | - | 3 - 4 | 333 | 97 |