The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004304.4 | NP_004295.2 | 21 - 22 | 4403 | 77 |
GenBank mRNA | AB209477.4 | - | 21 - 22 | 3940 | 77 |
EU788003.1 | - | 21 - 22 | 4358 | 77 | |
EU788004.1 | - | 21 - 22 | 4358 | 77 | |
EU788005.1 | - | 21 - 22 | 4358 | 77 | |
EU788006.1 | - | 21 - 22 | 4358 | 77 | |
EU788007.1 | - | 21 - 22 | 4358 | 77 | |
EU788008.1 | - | 21 - 22 | 4358 | 77 | |
EU788009.1 | - | 21 - 22 | 4358 | 77 | |
EU788010.1 | - | 21 - 22 | 4358 | 77 | |
FJ838786.1 | - | 21 - 22 | 4358 | 77 | |
FJ838787.1 | - | 21 - 22 | 4358 | 77 | |
GU128153.1 | - | 21 - 22 | 3451 | 77 | |
GU128154.1 | - | 21 - 22 | 3451 | 77 | |
GU128155.1 | - | 21 - 22 | 3451 | 77 | |
GU128156.1 | - | 21 - 22 | 3451 | 77 | |
GU128157.1 | - | 21 - 22 | 3451 | 77 | |
LN864494.1 | - | 2 - 3 | 651 | 77 | |
U62540.1 | - | 21 - 22 | 4362 | 77 | |
U66559.1 | - | 21 - 22 | 3599 | 77 |