The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002158.3 | NP_002149.2 | 3 - 4 | 849 | 110 |
XM_005264282.2 | XP_005264339.1 | 2 - 3 | 723 | 110 | |
XM_005264283.2 | XP_005264340.1 | 4 - 5 | 1044 | 110 | |
XM_006712001.3 | XP_006712064.1 | 3 - 4 | 780 | 110 | |
XM_006712002.3 | XP_006712065.1 | 2 - 3 | 740 | 110 | |
XM_011532808.2 | XP_011531110.1 | 4 - 5 | 966 | 110 | |
XM_017003956.1 | XP_016859445.1 | 3 - 4 | 1184 | 110 | |
XM_017003957.1 | XP_016859446.1 | 4 - 5 | 1041 | 110 | |
XM_017003958.1 | XP_016859447.1 | 4 - 5 | 1040 | 110 | |
XM_017003959.1 | XP_016859448.1 | 4 - 5 | 887 | 110 | |
GenBank mRNA | BC063305.1 | - | 3 - 4 | 774 | 110 |
BC069731.1 | - | 1 - 2 | 342 | 110 | |
U57029.1 | - | 1 - 2 | 545 | 110 |