Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA6
Entrez Gene ID
8973
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Gene Aliases
CHNRA6
Chromosome Location
Chr.8: 42752620 - 42768786 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004198.3 NP_004189.1 3 - 4 613 84
GenBank mRNA AK313521.1 - 3 - 4 459 84
BC014456.1 - 3 - 4 303 84
U62435.1 - 3 - 4 399 84
Y16282.1 - 3 - 4 257 84

View Full Product Details