The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171166.1 | NP_001164637.1 | 17 - 18 | 3232 | 53 |
NM_001171167.1 | NP_001164638.1 | 18 - 19 | 3671 | 53 | |
NM_001171168.1 | NP_001164639.1 | 17 - 18 | 3599 | 53 | |
NM_015099.3 | NP_055914.2 | 18 - 19 | 3327 | 53 | |
XM_006721478.3 | XP_006721541.1 | 18 - 19 | 3974 | 53 | |
XM_006721481.3 | XP_006721544.1 | 17 - 18 | 3887 | 53 | |
XM_006721482.3 | XP_006721545.1 | 16 - 17 | 3020 | 53 | |
XM_011523746.2 | XP_011522048.1 | 18 - 19 | 3334 | 53 | |
XM_011523747.2 | XP_011522049.1 | 18 - 19 | 3318 | 53 | |
XM_011523748.2 | XP_011522050.1 | 18 - 19 | 3333 | 53 | |
XM_011523749.2 | XP_011522051.1 | 18 - 19 | 3317 | 53 | |
GenBank mRNA | AB020716.1 | - | 18 - 19 | 3288 | 53 |
AL831849.1 | - | 17 - 18 | 3599 | 53 | |
AL833974.1 | - | 17 - 18 | 3217 | 53 | |
BC010050.2 | - | 10 - 11 | 1740 | 53 | |
BC016163.1 | - | 5 - 6 | 946 | 53 | |
BC032381.1 | - | 2 - 3 | 342 | 53 | |
BC136534.1 | - | 17 - 18 | 3256 | 53 | |
BC142695.1 | - | 18 - 19 | 3318 | 53 | |
BC144232.1 | - | 16 - 17 | 3184 | 53 | |
BC167148.1 | - | 18 - 19 | 3288 | 53 | |
BC167149.1 | - | 18 - 19 | 3288 | 53 | |
BC167151.1 | - | 18 - 19 | 3288 | 53 | |
BC167160.1 | - | 18 - 19 | 3288 | 53 |