The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171166.1 | NP_001164637.1 | 20 - 21 | 3572 | 117 |
NM_001171167.1 | NP_001164638.1 | 22 - 23 | 4030 | 117 | |
NM_001171168.1 | NP_001164639.1 | 21 - 22 | 3960 | 117 | |
NM_015099.3 | NP_055914.2 | 22 - 23 | 3688 | 117 | |
XM_006721478.3 | XP_006721541.1 | 22 - 23 | 4335 | 117 | |
XM_006721481.3 | XP_006721544.1 | 21 - 22 | 4248 | 117 | |
XM_006721482.3 | XP_006721545.1 | 20 - 21 | 3381 | 117 | |
XM_011523746.2 | XP_011522048.1 | 22 - 23 | 3695 | 117 | |
XM_011523747.2 | XP_011522049.1 | 22 - 23 | 3679 | 117 | |
XM_011523748.2 | XP_011522050.1 | 21 - 22 | 3673 | 117 | |
XM_011523749.2 | XP_011522051.1 | 21 - 22 | 3657 | 117 | |
GenBank mRNA | AB020716.1 | - | 22 - 23 | 3649 | 117 |
AL831849.1 | - | 21 - 22 | 3960 | 117 | |
BC010050.2 | - | 13 - 14 | 2080 | 117 | |
BC016163.1 | - | 9 - 10 | 1307 | 117 | |
BC032381.1 | - | 6 - 7 | 703 | 117 | |
BC136534.1 | - | 21 - 22 | 3617 | 117 | |
BC142695.1 | - | 22 - 23 | 3679 | 117 | |
BC144232.1 | - | 20 - 21 | 3545 | 117 | |
BC167148.1 | - | 22 - 23 | 3649 | 117 | |
BC167149.1 | - | 22 - 23 | 3649 | 117 | |
BC167151.1 | - | 22 - 23 | 3649 | 117 | |
BC167160.1 | - | 22 - 23 | 3649 | 117 |