The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171166.1 | NP_001164637.1 | 3 - 4 | 243 | 67 |
NM_001171167.1 | NP_001164638.1 | 4 - 5 | 682 | 67 | |
NM_001171168.1 | NP_001164639.1 | 4 - 5 | 682 | 67 | |
NM_015099.3 | NP_055914.2 | 4 - 5 | 338 | 67 | |
XM_006721478.3 | XP_006721541.1 | 4 - 5 | 970 | 67 | |
XM_006721481.3 | XP_006721544.1 | 4 - 5 | 970 | 67 | |
XM_006721482.3 | XP_006721545.1 | 3 - 4 | 192 | 67 | |
XM_011523746.2 | XP_011522048.1 | 4 - 5 | 330 | 67 | |
XM_011523747.2 | XP_011522049.1 | 4 - 5 | 329 | 67 | |
XM_011523748.2 | XP_011522050.1 | 4 - 5 | 329 | 67 | |
XM_011523749.2 | XP_011522051.1 | 4 - 5 | 328 | 67 | |
GenBank mRNA | AB020716.1 | - | 4 - 5 | 299 | 67 |
AK302528.1 | - | 4 - 5 | 210 | 67 | |
AL831849.1 | - | 4 - 5 | 682 | 67 | |
AL833974.1 | - | 3 - 4 | 228 | 67 | |
BC136534.1 | - | 3 - 4 | 267 | 67 | |
BC142695.1 | - | 4 - 5 | 329 | 67 | |
BC144232.1 | - | 3 - 4 | 267 | 67 | |
BC167148.1 | - | 4 - 5 | 299 | 67 | |
BC167149.1 | - | 4 - 5 | 299 | 67 | |
BC167151.1 | - | 4 - 5 | 299 | 67 | |
BC167160.1 | - | 4 - 5 | 299 | 67 | |
BI548947.1 | - | 4 - 5 | 332 | 67 | |
DB275145.1 | - | 3 - 4 | 244 | 67 |