The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145262.3 | NP_660305.2 | 3 - 4 | 624 | 100 |
NR_026700.1 | 2 - 3 | 208 | 100 | ||
NR_026701.1 | 3 - 4 | 624 | 100 | ||
XM_017005730.1 | XP_016861219.1 | 2 - 3 | 470 | 100 | |
GenBank mRNA | AF448855.1 | - | 3 - 4 | 625 | 100 |
AK074215.1 | - | 2 - 3 | 193 | 100 | |
BC042151.1 | - | 3 - 4 | 588 | 100 | |
DQ352863.1 | - | 2 - 3 | 528 | 100 |