Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16226342-16226609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
UniGene
Hs.71040
Gene Chromosome Location
Chr.19: 16197854-16235345 on Build GRCh38
Transcripts
NM_001130524.1 NM_032493.3 XM_017027420.1 XM_017027421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601556 c.602G>A p.R201Q Chr.19: 16226476-16226476 on Build GRCh38
4840048 c.693C>A p.L231L Chr.19: 16226531-16226531 on Build GRCh38
5981475 c.661G>A p.E221K Chr.19: 16226499-16226499 on Build GRCh38
4840049 c.657C>A p.L219L Chr.19: 16226531-16226531 on Build GRCh38
166438 c.576C>G p.S192R Chr.19: 16226450-16226450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961274615 Chr.19: 16226588-16226588 on Build GRCh38
rs914153321 Chr.19: 16226416-16226416 on Build GRCh38
rs756672040 Chr.19: 16226585-16226585 on Build GRCh38
rs774557887 Chr.19: 16226475-16226475 on Build GRCh38
rs775804469 Chr.19: 16226391-16226391 on Build GRCh38
rs777256405 Chr.19: 16226560-16226560 on Build GRCh38
rs149921079 Chr.19: 16226489-16226489 on Build GRCh38
rs770189406 Chr.19: 16226389-16226389 on Build GRCh38
rs948520653 Chr.19: 16226387-16226387 on Build GRCh38
rs764205073 Chr.19: 16226400-16226400 on Build GRCh38
rs767905458 Chr.19: 16226403-16226403 on Build GRCh38
rs1015628722 Chr.19: 16226576-16226576 on Build GRCh38
rs768123276 Chr.19: 16226498-16226498 on Build GRCh38
rs997387577 Chr.19: 16226436-16226436 on Build GRCh38
rs377436209 Chr.19: 16226470-16226470 on Build GRCh38
rs866359375 Chr.19: 16226464-16226465 on Build GRCh38
rs953913752 Chr.19: 16226383-16226383 on Build GRCh38
rs192837719 Chr.19: 16226568-16226568 on Build GRCh38
rs769670450 Chr.19: 16226451-16226451 on Build GRCh38
rs745679023 Chr.19: 16226446-16226446 on Build GRCh38
rs149668778 Chr.19: 16226432-16226432 on Build GRCh38
rs759220384 Chr.19: 16226384-16226384 on Build GRCh38
rs201947118 Chr.19: 16226402-16226402 on Build GRCh38
rs777141997 Chr.19: 16226427-16226427 on Build GRCh38
rs139438056 Chr.19: 16226409-16226409 on Build GRCh38
rs372660140 Chr.19: 16226410-16226410 on Build GRCh38
rs750850455 Chr.19: 16226519-16226519 on Build GRCh38
rs758109463 Chr.19: 16226558-16226558 on Build GRCh38
rs757965926 Chr.19: 16226426-16226426 on Build GRCh38
rs770937090 Chr.19: 16226437-16226437 on Build GRCh38
rs947084626 Chr.19: 16226411-16226411 on Build GRCh38
rs1005555130 Chr.19: 16226563-16226563 on Build GRCh38
rs147795014 Chr.19: 16226543-16226543 on Build GRCh38
rs752021520 Chr.19: 16226401-16226401 on Build GRCh38
rs746904475 Chr.19: 16226379-16226379 on Build GRCh38
rs780968690 Chr.19: 16226445-16226445 on Build GRCh38
rs150789905 Chr.19: 16226433-16226433 on Build GRCh38
rs920042774 Chr.19: 16226386-16226386 on Build GRCh38
rs776551598 Chr.19: 16226382-16226382 on Build GRCh38
rs752268709 Chr.19: 16226414-16226414 on Build GRCh38
rs139150489 Chr.19: 16226456-16226456 on Build GRCh38
rs763154481 Chr.19: 16226398-16226398 on Build GRCh38
rs762275752 Chr.19: 16226379-16226384 on Build GRCh38
rs763211790 Chr.19: 16226457-16226457 on Build GRCh38
rs761062270 Chr.19: 16226535-16226535 on Build GRCh38
rs922458808 Chr.19: 16226365-16226365 on Build GRCh38
rs770756827 Chr.19: 16226381-16226381 on Build GRCh38
rs750844005 Chr.19: 16226406-16226406 on Build GRCh38
rs752308670 Chr.19: 16226428-16226428 on Build GRCh38

View Full Product Details