Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52316387-52316660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT83
Entrez Gene ID
3889
Gene Name
keratin 83
Gene Aliases
HB3, Hb-3, KRTHB3
UniGene
Hs.720768
Gene Chromosome Location
Chr.12: 52314301-52321398 on Build GRCh38
Transcripts
NM_002282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
147522 c.1041+6A>G p.? Chr.12: 52316462-52316462 on Build GRCh38
7284690 c.916-3C>T p.? Chr.12: 52316596-52316596 on Build GRCh38
3812308 c.949G>A p.G317R Chr.12: 52316560-52316560 on Build GRCh38
6698281 c.965G>A p.R322H Chr.12: 52316544-52316544 on Build GRCh38
1747066 c.939G>A p.V313V Chr.12: 52316570-52316570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766601032 Chr.12: 52316531-52316533 on Build GRCh38
rs774321223 Chr.12: 52316521-52316521 on Build GRCh38
rs371272186 Chr.12: 52316619-52316619 on Build GRCh38
rs765443130 Chr.12: 52316538-52316538 on Build GRCh38
rs759728232 Chr.12: 52316634-52316635 on Build GRCh38
rs749276155 Chr.12: 52316515-52316515 on Build GRCh38
rs763011771 Chr.12: 52316623-52316623 on Build GRCh38
rs777009664 Chr.12: 52316536-52316536 on Build GRCh38
rs573181731 Chr.12: 52316415-52316415 on Build GRCh38
rs754111776 Chr.12: 52316464-52316464 on Build GRCh38
rs752671222 Chr.12: 52316542-52316542 on Build GRCh38
rs762240456 Chr.12: 52316631-52316631 on Build GRCh38
rs745505440 Chr.12: 52316496-52316496 on Build GRCh38
rs752019274 Chr.12: 52316629-52316629 on Build GRCh38
rs553152302 Chr.12: 52316491-52316491 on Build GRCh38
rs996735954 Chr.12: 52316584-52316584 on Build GRCh38
rs55722266 Chr.12: 52316414-52316414 on Build GRCh38
rs561325506 Chr.12: 52316576-52316576 on Build GRCh38
rs138807826 Chr.12: 52316564-52316564 on Build GRCh38
rs145387028 Chr.12: 52316545-52316545 on Build GRCh38
rs779634636 Chr.12: 52316560-52316560 on Build GRCh38
rs755719932 Chr.12: 52316552-52316552 on Build GRCh38
rs768424588 Chr.12: 52316519-52316519 on Build GRCh38
rs371181908 Chr.12: 52316447-52316447 on Build GRCh38
rs534990362 Chr.12: 52316587-52316587 on Build GRCh38
rs776245337 Chr.12: 52316434-52316434 on Build GRCh38
rs767410937 Chr.12: 52316529-52316529 on Build GRCh38
rs748796258 Chr.12: 52316561-52316561 on Build GRCh38
rs773105273 Chr.12: 52316577-52316577 on Build GRCh38
rs746706438 Chr.12: 52316431-52316431 on Build GRCh38
rs146734638 Chr.12: 52316514-52316514 on Build GRCh38
rs777933385 Chr.12: 52316424-52316424 on Build GRCh38
rs201828918 Chr.12: 52316547-52316547 on Build GRCh38
rs753256333 Chr.12: 52316488-52316488 on Build GRCh38
rs201756344 Chr.12: 52316456-52316456 on Build GRCh38
rs758485277 Chr.12: 52316544-52316544 on Build GRCh38
rs1053874619 Chr.12: 52316446-52316446 on Build GRCh38
rs759085887 Chr.12: 52316441-52316441 on Build GRCh38
rs748042840 Chr.12: 52316570-52316570 on Build GRCh38
rs764219879 Chr.12: 52316628-52316628 on Build GRCh38
rs781477861 Chr.12: 52316549-52316549 on Build GRCh38
rs2257286 Chr.12: 52316525-52316525 on Build GRCh38
rs370476740 Chr.12: 52316459-52316459 on Build GRCh38
rs143467763 Chr.12: 52316492-52316492 on Build GRCh38
rs923979287 Chr.12: 52316635-52316635 on Build GRCh38
rs2270267 Chr.12: 52316462-52316462 on Build GRCh38
rs769306649 Chr.12: 52316510-52316510 on Build GRCh38
rs199773372 Chr.12: 52316455-52316455 on Build GRCh38
rs770443869 Chr.12: 52316433-52316433 on Build GRCh38
rs200128355 Chr.12: 52316548-52316548 on Build GRCh38
rs745523832 Chr.12: 52316550-52316550 on Build GRCh38
rs377435485 Chr.12: 52316463-52316463 on Build GRCh38
rs769287971 Chr.12: 52316449-52316449 on Build GRCh38
rs763111001 Chr.12: 52316475-52316476 on Build GRCh38
rs774347479 Chr.12: 52316587-52316588 on Build GRCh38
rs202206430 Chr.12: 52316495-52316495 on Build GRCh38
rs765354005 Chr.12: 52316486-52316486 on Build GRCh38
rs755169515 Chr.12: 52316465-52316465 on Build GRCh38
rs775463584 Chr.12: 52316451-52316451 on Build GRCh38
rs758792997 Chr.12: 52316489-52316489 on Build GRCh38

View Full Product Details