Product Details

Assay Reference Genome
Location

Chr.2:27367118 on build GRCh38
Cytoband
2p23.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8890
Gene Symbol
EIF2B4
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
Location
Chr.2:27364352-27370457 on build GRCh38
Assay Gene Location
Overlaps Intron 10 - Exon 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
EIF2B4 NM_001034116.1 NP_001029288.1
NM_001318965.1 NP_001305894.1
NM_001318966.1 NP_001305895.1
NM_001318967.1 NP_001305896.1
NM_001318968.1 NP_001305897.1
NM_001318969.1 NP_001305898.1
NM_015636.3 NP_056451.3
NM_172195.3 NP_751945.2
XM_006712132.1 XP_006712195.1
XM_011533147.2 XP_011531449.1
AB209532.1 3 2730 BAD92769.1
AF057699.1 AAQ13438.1
AF086520.1
AF112207.1 AAF17195.1
AF218009.1 AAG17251.1
AJ011305.1 CAB57260.1
AJ011306.1 CAB57261.1
AK300734.1
AL050109.1 CAB43277.1
BC001870.1 AAH01870.1
BC091502.1 AAH91502.1
JQ991007.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833614 Chr2:27263559 - 27452213 on Build GRCh38 Gain UCN ZNF513 PPM1G LOC105374363 IFT172 MPV17 TRIM54 GTF3C2-AS1 SNX17 KRTCAP3 FTH1P3 NRBP1 SLC30A3 DNAJC5G EIF2B4 GTF3C2
esv3575209 Chr2:27334578 - 27389381 on Build GRCh38 Gain GTF3C2-AS1 SNX17 ZNF513 PPM1G EIF2B4 LOC105374363 GTF3C2
nsv581244 Chr2:27335385 - 27480628 on Build GRCh38 Loss GTF3C2-AS1 SNX17 KRTCAP3 FTH1P3 NRBP1 ZNF513 PPM1G EIF2B4 LOC105374363 GTF3C2 IFT172

View Full Product Details