Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGR6
Entrez Gene ID
59352
Gene Name
leucine rich repeat containing G protein-coupled receptor 6
Gene Aliases
GPCR, VTS20631
Chromosome Location
Chr.1: 202193990 - 202319761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017403.1 NP_001017403.1 2 - 3 286 59
NM_001017404.1 NP_001017404.1 2 - 3 157 59
NM_021636.2 NP_067649.2 2 - 3 173 59
XM_005245404.3 XP_005245461.1 2 - 3 230 59
XM_011509838.2 XP_011508140.1 2 - 3 231 59
XM_011509839.2 XP_011508141.1 2 - 3 232 59
XM_011509840.2 XP_011508142.1 2 - 3 231 59
XM_017001996.1 XP_016857485.1 2 - 3 232 59
GenBank mRNA AB049405.1 - 2 - 3 171 59
AF190501.1 - 2 - 3 157 59
AY358119.1 - 2 - 3 214 59
BC047905.1 - 2 - 3 156 59

View Full Product Details