Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
Chromosome Location
Chr.11: 64606174 - 64723188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015080.3 NP_055895.1 7 - 8 1659 93
XM_011545370.1 XP_011543672.1 6 - 7 1212 93
XM_011545371.1 XP_011543673.1 6 - 7 1209 93
XM_017018563.1 XP_016874052.1 5 - 6 1182 93
XM_017018564.1 XP_016874053.1 5 - 6 1179 93
XM_017018565.1 XP_016874054.1 6 - 7 1212 93
XM_017018566.1 XP_016874055.1 5 - 6 1140 93
XM_017018567.1 XP_016874056.1 6 - 7 1212 93
XM_017018568.1 XP_016874057.1 6 - 7 1212 93
XM_017018569.1 XP_016874058.1 4 - 5 1110 93
XM_017018570.1 XP_016874059.1 6 - 7 1212 93
XM_017018571.1 XP_016874060.1 6 - 7 1212 93
XM_017018572.1 XP_016874061.1 6 - 7 507 93
XM_017018573.1 XP_016874062.1 6 - 7 1212 93
XM_017018574.1 XP_016874063.1 5 - 6 477 93
XM_017018575.1 XP_016874064.1 5 - 6 474 93
XM_017018578.1 XP_016874067.1 6 - 7 1212 93
GenBank mRNA AB035266.1 - 6 - 7 1311 93

View Full Product Details