Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40394310-40394547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
UniGene
Hs.205457
Gene Chromosome Location
Chr.19: 40393762-40414718 on Build GRCh38
Transcripts
NM_020956.2 NM_181882.2 XM_011527171.2 XM_017027046.1 XM_017027047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
314508 c.3921G>T p.R1307R Chr.19: 40394431-40394431 on Build GRCh38
5743993 c.4004G>A p.R1335Q Chr.19: 40394348-40394348 on Build GRCh38
4923225 c.3875A>T p.Q1292L Chr.19: 40394477-40394477 on Build GRCh38
246822 c.4001C>T p.P1334L Chr.19: 40394351-40394351 on Build GRCh38
4078212 c.3845C>T p.S1282L Chr.19: 40394507-40394507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753530396 Chr.19: 40394507-40394507 on Build GRCh38
rs777921155 Chr.19: 40394447-40394447 on Build GRCh38
rs779070838 Chr.19: 40394453-40394453 on Build GRCh38
rs948370389 Chr.19: 40394414-40394414 on Build GRCh38
rs759245840 Chr.19: 40394485-40394485 on Build GRCh38
rs370802101 Chr.19: 40394526-40394526 on Build GRCh38
rs757223803 Chr.19: 40394452-40394452 on Build GRCh38
rs200139733 Chr.19: 40394347-40394347 on Build GRCh38
rs886188493 Chr.19: 40394501-40394501 on Build GRCh38
rs140109585 Chr.19: 40394413-40394413 on Build GRCh38
rs751126995 Chr.19: 40394424-40394424 on Build GRCh38
rs779565946 Chr.19: 40394344-40394344 on Build GRCh38
rs901951321 Chr.19: 40394455-40394455 on Build GRCh38
rs551830109 Chr.19: 40394423-40394423 on Build GRCh38
rs747247972 Chr.19: 40394517-40394517 on Build GRCh38
rs774367348 Chr.19: 40394406-40394406 on Build GRCh38
rs768961174 Chr.19: 40394518-40394518 on Build GRCh38
rs763990676 Chr.19: 40394498-40394498 on Build GRCh38
rs754692947 Chr.19: 40394427-40394427 on Build GRCh38
rs143289108 Chr.19: 40394506-40394506 on Build GRCh38
rs757247849 Chr.19: 40394503-40394503 on Build GRCh38
rs149626915 Chr.19: 40394392-40394392 on Build GRCh38
rs769014032 Chr.19: 40394466-40394466 on Build GRCh38
rs752593001 Chr.19: 40394433-40394433 on Build GRCh38
rs756103463 Chr.19: 40394446-40394446 on Build GRCh38
rs776947855 Chr.19: 40394467-40394467 on Build GRCh38
rs374370759 Chr.19: 40394499-40394499 on Build GRCh38
rs942931158 Chr.19: 40394470-40394470 on Build GRCh38
rs527265816 Chr.19: 40394343-40394343 on Build GRCh38
rs146205352 Chr.19: 40394514-40394514 on Build GRCh38
rs142762689 Chr.19: 40394405-40394405 on Build GRCh38
rs377639106 Chr.19: 40394520-40394520 on Build GRCh38
rs749381927 Chr.19: 40394404-40394404 on Build GRCh38
rs773955274 Chr.19: 40394484-40394484 on Build GRCh38
rs532367972 Chr.19: 40394412-40394412 on Build GRCh38
rs749975234 Chr.19: 40394333-40394333 on Build GRCh38
rs764644013 Chr.19: 40394417-40394417 on Build GRCh38
rs887982096 Chr.19: 40394437-40394437 on Build GRCh38
rs555523161 Chr.19: 40394496-40394496 on Build GRCh38
rs775809057 Chr.19: 40394461-40394461 on Build GRCh38
rs369372933 Chr.19: 40394509-40394509 on Build GRCh38
rs762407223 Chr.19: 40394468-40394468 on Build GRCh38
rs749251511 Chr.19: 40394451-40394451 on Build GRCh38
rs772705019 Chr.19: 40394422-40394422 on Build GRCh38
rs781159482 Chr.19: 40394389-40394389 on Build GRCh38
rs767394584 Chr.19: 40394432-40394432 on Build GRCh38
rs748063094 Chr.19: 40394391-40394391 on Build GRCh38
rs772371124 Chr.19: 40394409-40394409 on Build GRCh38
rs534186199 Chr.19: 40394472-40394472 on Build GRCh38
rs758057597 Chr.19: 40394335-40394335 on Build GRCh38
rs752531823 Chr.19: 40394486-40394486 on Build GRCh38
rs202219076 Chr.19: 40394458-40394458 on Build GRCh38
rs957373519 Chr.19: 40394421-40394421 on Build GRCh38
rs567324732 Chr.19: 40394500-40394500 on Build GRCh38
rs747442188 Chr.19: 40394465-40394465 on Build GRCh38
rs765769524 Chr.19: 40394479-40394479 on Build GRCh38
rs777974539 Chr.19: 40394398-40394398 on Build GRCh38
rs185914183 Chr.19: 40394408-40394408 on Build GRCh38
rs559968504 Chr.19: 40394349-40394349 on Build GRCh38
rs754673395 Chr.19: 40394354-40394354 on Build GRCh38
rs577796628 Chr.19: 40394521-40394521 on Build GRCh38

View Full Product Details