The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175881.3 | NP_787077.1 | 2 - 3 | 452 | 106 |
GenBank mRNA | BC039862.1 | - | 2 - 3 | 452 | 106 |