Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERINC5
Entrez Gene ID
256987
Gene Name
serine incorporator 5
Gene Aliases
C5orf12, TPO1
Chromosome Location
Chr.5: 80111227 - 80256082 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174071.2 NP_001167542.1 5 - 6 713 71
NM_001174072.2 NP_001167543.1 5 - 6 713 71
NM_178276.6 NP_840060.1 5 - 6 713 71
NR_126060.1 5 - 6 713 71
NR_126061.1 5 - 6 713 71
XM_011543304.1 XP_011541606.1 5 - 6 678 71
XM_017009323.1 XP_016864812.1 5 - 6 659 71
XM_017009324.1 XP_016864813.1 5 - 6 678 71
GenBank mRNA AF498273.1 - 5 - 6 683 71
AK297467.1 - 5 - 6 710 71
BC101280.2 - 5 - 6 645 71
BC101281.3 - 5 - 6 650 71
BC101283.2 - 5 - 6 650 71

View Full Product Details