Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46031471-46031732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620001 c.3220C>T p.R1074W Chr.17: 46031574-46031574 on Build GRCh38
139063 c.3271C>T p.R1091W Chr.17: 46031523-46031523 on Build GRCh38
6690061 c.3171G>T p.Q1057H Chr.17: 46031623-46031623 on Build GRCh38
132716 c.3113G>A p.R1038Q Chr.17: 46031681-46031681 on Build GRCh38
158679 c.3237G>A p.A1079A Chr.17: 46031557-46031557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767917046 Chr.17: 46031700-46031700 on Build GRCh38
rs370112084 Chr.17: 46031558-46031558 on Build GRCh38
rs753191139 Chr.17: 46031495-46031495 on Build GRCh38
rs764750830 Chr.17: 46031576-46031576 on Build GRCh38
rs201083879 Chr.17: 46031624-46031624 on Build GRCh38
rs747976314 Chr.17: 46031557-46031557 on Build GRCh38
rs762231138 Chr.17: 46031608-46031608 on Build GRCh38
rs769463549 Chr.17: 46031667-46031667 on Build GRCh38
rs922454072 Chr.17: 46031523-46031523 on Build GRCh38
rs770548045 Chr.17: 46031655-46031655 on Build GRCh38
rs34579536 Chr.17: 46031540-46031540 on Build GRCh38
rs760084513 Chr.17: 46031704-46031705 on Build GRCh38
rs796052590 Chr.17: 46031646-46031646 on Build GRCh38
rs760256746 Chr.17: 46031564-46031564 on Build GRCh38
rs762427291 Chr.17: 46031684-46031684 on Build GRCh38
rs200719065 Chr.17: 46031651-46031651 on Build GRCh38
rs960036500 Chr.17: 46031671-46031671 on Build GRCh38
rs368475798 Chr.17: 46031597-46031597 on Build GRCh38
rs746671618 Chr.17: 46031654-46031654 on Build GRCh38
rs989483142 Chr.17: 46031705-46031705 on Build GRCh38
rs982701618 Chr.17: 46031604-46031604 on Build GRCh38
rs776183957 Chr.17: 46031656-46031656 on Build GRCh38
rs764865498 Chr.17: 46031504-46031506 on Build GRCh38
rs776071732 Chr.17: 46031570-46031570 on Build GRCh38
rs771796347 Chr.17: 46031648-46031648 on Build GRCh38
rs797045049 Chr.17: 46031669-46031669 on Build GRCh38
rs778781627 Chr.17: 46031635-46031635 on Build GRCh38
rs763541067 Chr.17: 46031573-46031573 on Build GRCh38
rs374360315 Chr.17: 46031706-46031706 on Build GRCh38
rs1001573190 Chr.17: 46031559-46031559 on Build GRCh38
rs750644383 Chr.17: 46031536-46031536 on Build GRCh38
rs781609742 Chr.17: 46031534-46031534 on Build GRCh38
rs779999524 Chr.17: 46031548-46031548 on Build GRCh38
rs768018906 Chr.17: 46031609-46031609 on Build GRCh38
rs772894204 Chr.17: 46031650-46031651 on Build GRCh38
rs140670092 Chr.17: 46031665-46031665 on Build GRCh38
rs796052598 Chr.17: 46031496-46031496 on Build GRCh38
rs770409760 Chr.17: 46031569-46031569 on Build GRCh38
rs774951690 Chr.17: 46031677-46031677 on Build GRCh38
rs942015604 Chr.17: 46031631-46031631 on Build GRCh38
rs560719955 Chr.17: 46031499-46031499 on Build GRCh38
rs529738082 Chr.17: 46031507-46031507 on Build GRCh38
rs1037675968 Chr.17: 46031640-46031640 on Build GRCh38
rs756355714 Chr.17: 46031617-46031617 on Build GRCh38
rs372299492 Chr.17: 46031626-46031626 on Build GRCh38
rs752029022 Chr.17: 46031522-46031522 on Build GRCh38
rs765828427 Chr.17: 46031494-46031494 on Build GRCh38
rs780165589 Chr.17: 46031618-46031618 on Build GRCh38
rs199769766 Chr.17: 46031556-46031556 on Build GRCh38
rs370904425 Chr.17: 46031530-46031530 on Build GRCh38
rs796052589 Chr.17: 46031653-46031653 on Build GRCh38
rs768634007 Chr.17: 46031555-46031555 on Build GRCh38
rs749241540 Chr.17: 46031549-46031549 on Build GRCh38
rs746504988 Chr.17: 46031650-46031650 on Build GRCh38
rs542543248 Chr.17: 46031489-46031489 on Build GRCh38
rs748104970 Chr.17: 46031647-46031647 on Build GRCh38
rs772870153 Chr.17: 46031561-46031561 on Build GRCh38
rs750589073 Chr.17: 46031613-46031613 on Build GRCh38
rs963795454 Chr.17: 46031519-46031519 on Build GRCh38

View Full Product Details