The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182511.3 | NP_872317.1 | 3 - 4 | 1075 | 76 |
XM_006722394.3 | XP_006722457.1 | 5 - 6 | 1422 | 76 | |
XM_011525824.2 | XP_011524126.1 | 5 - 6 | 1425 | 76 | |
XM_017025559.1 | XP_016881048.1 | 5 - 6 | 1926 | 76 | |
XM_017025560.1 | XP_016881049.1 | 2 - 3 | 587 | 76 | |
GenBank mRNA | AK291364.1 | - | 2 - 3 | 587 | 76 |
AY359074.1 | - | 3 - 4 | 915 | 76 | |
BC035789.1 | - | 2 - 3 | 577 | 76 |