Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 13921858-13922131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
UniGene
Hs.567265
Gene Chromosome Location
Chr.16: 13920138-13952348 on Build GRCh38
Transcripts
NM_005236.2 XM_011522424.2 XM_017023043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6143489 c.290G>T p.R97L Chr.16: 13922113-13922113 on Build GRCh38
6974459 c.233T>C p.I78T Chr.16: 13922056-13922056 on Build GRCh38
4823334 c.208G>A p.E70K Chr.16: 13922031-13922031 on Build GRCh38
6921482 c.257G>A p.R86H Chr.16: 13922080-13922080 on Build GRCh38
1629778 c.307C>G p.Q103E Chr.16: 13922130-13922130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771203473 Chr.16: 13922057-13922059 on Build GRCh38
rs769932063 Chr.16: 13922079-13922079 on Build GRCh38
rs1010878897 Chr.16: 13921953-13921953 on Build GRCh38
rs55761944 Chr.16: 13922064-13922064 on Build GRCh38
rs759518166 Chr.16: 13922031-13922031 on Build GRCh38
rs756240191 Chr.16: 13921919-13921921 on Build GRCh38
rs1014546174 Chr.16: 13922071-13922071 on Build GRCh38
rs889499753 Chr.16: 13921913-13921913 on Build GRCh38
rs750687183 Chr.16: 13922062-13922062 on Build GRCh38
rs192488568 Chr.16: 13922048-13922048 on Build GRCh38
rs139142361 Chr.16: 13921997-13921997 on Build GRCh38
rs749125796 Chr.16: 13922015-13922015 on Build GRCh38
rs371487368 Chr.16: 13922083-13922083 on Build GRCh38
rs556330628 Chr.16: 13922098-13922098 on Build GRCh38
rs892488883 Chr.16: 13921939-13921939 on Build GRCh38
rs200805983 Chr.16: 13921992-13921992 on Build GRCh38
rs771860466 Chr.16: 13922033-13922033 on Build GRCh38
rs141591400 Chr.16: 13922040-13922040 on Build GRCh38
rs750930128 Chr.16: 13921987-13921987 on Build GRCh38
rs751713894 Chr.16: 13922068-13922068 on Build GRCh38
rs3136056 Chr.16: 13922075-13922075 on Build GRCh38
rs187435008 Chr.16: 13922080-13922080 on Build GRCh38
rs560264535 Chr.16: 13921911-13921911 on Build GRCh38
rs747024254 Chr.16: 13922001-13922001 on Build GRCh38
rs191886782 Chr.16: 13922082-13922082 on Build GRCh38
rs753534967 Chr.16: 13922049-13922049 on Build GRCh38
rs770696743 Chr.16: 13922016-13922016 on Build GRCh38
rs1035229051 Chr.16: 13922042-13922042 on Build GRCh38
rs748380872 Chr.16: 13922076-13922076 on Build GRCh38
rs781373518 Chr.16: 13922012-13922012 on Build GRCh38
rs773956647 Chr.16: 13922028-13922028 on Build GRCh38
rs768893462 Chr.16: 13922009-13922009 on Build GRCh38
rs145315496 Chr.16: 13922034-13922034 on Build GRCh38
rs765712858 Chr.16: 13922055-13922055 on Build GRCh38
rs755021656 Chr.16: 13922070-13922070 on Build GRCh38
rs61760162 Chr.16: 13922051-13922051 on Build GRCh38

View Full Product Details