Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56451961-56452186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
UniGene
Hs.473144
Gene Chromosome Location
Chr.20: 56411550-56460240 on Build GRCh38
Transcripts
NM_001164114.1 NM_001164115.1 NM_001164116.1 NM_020356.3 XM_006723831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174998 c.903G>A p.Q301Q Chr.20: 56452079-56452079 on Build GRCh38
6558905 c.890A>T p.N297I Chr.20: 56452066-56452066 on Build GRCh38
3548038 c.1004T>A p.F335Y Chr.20: 56452180-56452180 on Build GRCh38
6525970 c.1003T>A p.F335I Chr.20: 56452179-56452179 on Build GRCh38
6533123 c.979G>C p.V327L Chr.20: 56452155-56452155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749532978 Chr.20: 56452005-56452005 on Build GRCh38
rs773645729 Chr.20: 56452127-56452127 on Build GRCh38
rs143746875 Chr.20: 56451992-56451992 on Build GRCh38
rs376819122 Chr.20: 56452059-56452059 on Build GRCh38
rs757153716 Chr.20: 56452049-56452049 on Build GRCh38
rs779074764 Chr.20: 56452060-56452060 on Build GRCh38
rs369609263 Chr.20: 56452121-56452121 on Build GRCh38
rs771671673 Chr.20: 56451987-56451987 on Build GRCh38
rs746727406 Chr.20: 56452073-56452073 on Build GRCh38
rs748193269 Chr.20: 56452041-56452041 on Build GRCh38
rs753397576 Chr.20: 56452042-56452042 on Build GRCh38
rs780212117 Chr.20: 56451989-56451989 on Build GRCh38
rs754293038 Chr.20: 56452002-56452002 on Build GRCh38
rs1023591643 Chr.20: 56452139-56452139 on Build GRCh38
rs781391496 Chr.20: 56452039-56452039 on Build GRCh38
rs766918491 Chr.20: 56452134-56452134 on Build GRCh38
rs1012790504 Chr.20: 56452088-56452088 on Build GRCh38
rs140450802 Chr.20: 56452152-56452152 on Build GRCh38
rs765728156 Chr.20: 56452119-56452119 on Build GRCh38
rs772751260 Chr.20: 56452159-56452159 on Build GRCh38
rs780405743 Chr.20: 56452027-56452027 on Build GRCh38
rs763382815 Chr.20: 56452133-56452133 on Build GRCh38
rs775146098 Chr.20: 56451988-56451988 on Build GRCh38
rs956151734 Chr.20: 56452111-56452111 on Build GRCh38
rs763631609 Chr.20: 56451993-56451993 on Build GRCh38
rs778745360 Chr.20: 56452054-56452054 on Build GRCh38
rs1031311340 Chr.20: 56452103-56452103 on Build GRCh38
rs865932743 Chr.20: 56452140-56452140 on Build GRCh38
rs199808780 Chr.20: 56452021-56452021 on Build GRCh38
rs762438061 Chr.20: 56452018-56452018 on Build GRCh38
rs1022703221 Chr.20: 56452033-56452033 on Build GRCh38
rs745625249 Chr.20: 56452055-56452055 on Build GRCh38
rs776146546 Chr.20: 56452100-56452100 on Build GRCh38
rs369739655 Chr.20: 56452081-56452081 on Build GRCh38
rs200949271 Chr.20: 56452095-56452095 on Build GRCh38
rs999874366 Chr.20: 56452078-56452078 on Build GRCh38
rs758859344 Chr.20: 56452023-56452023 on Build GRCh38
rs372172040 Chr.20: 56452034-56452034 on Build GRCh38
rs761263769 Chr.20: 56452104-56452104 on Build GRCh38
rs531219703 Chr.20: 56452086-56452086 on Build GRCh38
rs1035946876 Chr.20: 56452017-56452017 on Build GRCh38
rs775091205 Chr.20: 56452066-56452066 on Build GRCh38

View Full Product Details