Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18787322-18787591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
UniGene
Hs.1584
Gene Chromosome Location
Chr.19: 18782773-18791305 on Build GRCh38
Transcripts
NM_000095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6270363 c.1068C>A p.N356K Chr.19: 18787558-18787558 on Build GRCh38
6633978 c.1068C>T p.N356N Chr.19: 18787558-18787558 on Build GRCh38
7167068 c.1036G>A p.D346N Chr.19: 18787590-18787590 on Build GRCh38
5855104 c.1135+1G>A p.? Chr.19: 18787490-18787490 on Build GRCh38
4750719 c.1109C>T p.A370V Chr.19: 18787517-18787517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772474421 Chr.19: 18787559-18787559 on Build GRCh38
rs765277476 Chr.19: 18787495-18787495 on Build GRCh38
rs887728307 Chr.19: 18787392-18787392 on Build GRCh38
rs998843596 Chr.19: 18787460-18787460 on Build GRCh38
rs373019491 Chr.19: 18787450-18787450 on Build GRCh38
rs891171265 Chr.19: 18787394-18787394 on Build GRCh38
rs760831704 Chr.19: 18787466-18787466 on Build GRCh38
rs780972997 Chr.19: 18787516-18787516 on Build GRCh38
rs895484907 Chr.19: 18787475-18787475 on Build GRCh38
rs983412693 Chr.19: 18787348-18787348 on Build GRCh38
rs562232083 Chr.19: 18787344-18787344 on Build GRCh38
rs113438423 Chr.19: 18787471-18787471 on Build GRCh38
rs750873429 Chr.19: 18787460-18787460 on Build GRCh38
rs779651201 Chr.19: 18787442-18787442 on Build GRCh38
rs773580745 Chr.19: 18787561-18787561 on Build GRCh38
rs751496694 Chr.19: 18787508-18787508 on Build GRCh38
rs755776825 Chr.19: 18787517-18787517 on Build GRCh38
rs779823081 Chr.19: 18787524-18787524 on Build GRCh38
rs753872206 Chr.19: 18787470-18787470 on Build GRCh38
rs142274526 Chr.19: 18787572-18787572 on Build GRCh38
rs748999247 Chr.19: 18787526-18787526 on Build GRCh38
rs1054969546 Chr.19: 18787419-18787419 on Build GRCh38
rs374900253 Chr.19: 18787480-18787480 on Build GRCh38
rs1008231904 Chr.19: 18787407-18787407 on Build GRCh38
rs1047659844 Chr.19: 18787376-18787376 on Build GRCh38
rs778354590 Chr.19: 18787528-18787528 on Build GRCh38
rs757625411 Chr.19: 18787443-18787443 on Build GRCh38
rs1031874618 Chr.19: 18787501-18787501 on Build GRCh38
rs957655517 Chr.19: 18787507-18787507 on Build GRCh38
rs1024288093 Chr.19: 18787432-18787432 on Build GRCh38
rs748840065 Chr.19: 18787447-18787447 on Build GRCh38
rs894526957 Chr.19: 18787420-18787420 on Build GRCh38
rs762285685 Chr.19: 18787463-18787463 on Build GRCh38
rs754522846 Chr.19: 18787527-18787527 on Build GRCh38
rs757207641 Chr.19: 18787509-18787509 on Build GRCh38
rs371522589 Chr.19: 18787439-18787439 on Build GRCh38
rs1022585347 Chr.19: 18787533-18787533 on Build GRCh38
rs748625870 Chr.19: 18787554-18787554 on Build GRCh38
rs531255842 Chr.19: 18787367-18787367 on Build GRCh38
rs369968743 Chr.19: 18787417-18787417 on Build GRCh38
rs771322282 Chr.19: 18787571-18787571 on Build GRCh38
rs866318237 Chr.19: 18787354-18787354 on Build GRCh38
rs772485716 Chr.19: 18787464-18787464 on Build GRCh38
rs922942417 Chr.19: 18787366-18787366 on Build GRCh38
rs921381347 Chr.19: 18787421-18787421 on Build GRCh38
rs138804934 Chr.19: 18787567-18787567 on Build GRCh38
rs1013432678 Chr.19: 18787422-18787422 on Build GRCh38
rs766613369 Chr.19: 18787467-18787467 on Build GRCh38
rs768004340 Chr.19: 18787451-18787456 on Build GRCh38
rs774901147 Chr.19: 18787456-18787456 on Build GRCh38
rs908561612 Chr.19: 18787353-18787353 on Build GRCh38

View Full Product Details