Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37850063-37850312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
303498 c.16C>T p.R6* Chr.7: 37850282-37850282 on Build GRCh38
3638513 c.25C>T p.Q9* Chr.7: 37850291-37850291 on Build GRCh38
1450586 c.9_10insA p.R6fs*11 Chr.7: 37850275-37850276 on Build GRCh38
6028283 c.10_11insA p.R6fs*11 Chr.7: 37850276-37850277 on Build GRCh38
3638512 c.17G>T p.R6L Chr.7: 37850283-37850283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750045501 Chr.7: 37850282-37850282 on Build GRCh38
rs184571218 Chr.7: 37850264-37850264 on Build GRCh38
rs771208086 Chr.7: 37850236-37850236 on Build GRCh38
rs202203190 Chr.7: 37850240-37850240 on Build GRCh38
rs200067292 Chr.7: 37850262-37850262 on Build GRCh38
rs141149150 Chr.7: 37850237-37850240 on Build GRCh38
rs765828426 Chr.7: 37850290-37850290 on Build GRCh38
rs765951801 Chr.7: 37850257-37850259 on Build GRCh38
rs373353542 Chr.7: 37850291-37850291 on Build GRCh38
rs781406524 Chr.7: 37850238-37850238 on Build GRCh38
rs765624306 Chr.7: 37850186-37850186 on Build GRCh38
rs769797120 Chr.7: 37850250-37850250 on Build GRCh38
rs995388681 Chr.7: 37850127-37850127 on Build GRCh38
rs899677193 Chr.7: 37850116-37850116 on Build GRCh38
rs879588920 Chr.7: 37850208-37850208 on Build GRCh38
rs367838558 Chr.7: 37850252-37850252 on Build GRCh38
rs1026906141 Chr.7: 37850167-37850167 on Build GRCh38
rs767216992 Chr.7: 37850269-37850269 on Build GRCh38
rs888972953 Chr.7: 37850100-37850100 on Build GRCh38
rs778157928 Chr.7: 37850226-37850226 on Build GRCh38
rs745894785 Chr.7: 37850241-37850241 on Build GRCh38
rs771112241 Chr.7: 37850251-37850251 on Build GRCh38
rs981731927 Chr.7: 37850278-37850278 on Build GRCh38
rs747201006 Chr.7: 37850234-37850234 on Build GRCh38
rs768545346 Chr.7: 37850260-37850260 on Build GRCh38
rs762850101 Chr.7: 37850258-37850258 on Build GRCh38
rs752091063 Chr.7: 37850163-37850163 on Build GRCh38
rs10271309 Chr.7: 37850184-37850184 on Build GRCh38
rs199576209 Chr.7: 37850283-37850283 on Build GRCh38
rs758711906 Chr.7: 37850222-37850222 on Build GRCh38
rs560280586 Chr.7: 37850244-37850247 on Build GRCh38

View Full Product Details