Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10orf54
Entrez Gene ID
64115
Gene Name
chromosome 10 open reading frame 54
Gene Aliases
B7-H5, B7H5, DD1alpha, GI24, PD-1H, PP2135, SISP1, VISTA
Chromosome Location
Chr.10: 71747556 - 71773580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022153.1 NP_071436.1 2 - 3 656 77
GenBank mRNA AB073653.1 - - 917 77
AF289594.1 - - 1092 77
AK024449.1 - 2 - 3 574 77
AY265805.1 - 2 - 3 571 77
AY358379.1 - 2 - 3 656 77
BC020568.1 - 2 - 3 528 77
BC111048.1 - 2 - 3 504 77
EF506490.1 - 2 - 3 568 77

View Full Product Details