Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
Chromosome Location
Chr.1: 149928651 - 149936898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145862.1 NP_001139334.1 12 - 13 1373 62
NM_181873.3 NP_870988.2 11 - 12 1115 62
XM_006711137.1 XP_006711200.1 11 - 12 1092 62
GenBank mRNA AK097000.1 - 11 - 12 1092 62
AK298775.1 - 12 - 13 1342 62
BC100818.2 - 11 - 12 1033 62
BC100819.2 - 11 - 12 1033 62
BC100820.1 - 11 - 12 1033 62

View Full Product Details