Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
Chromosome Location
Chr.1: 149928651 - 149936898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145862.1 NP_001139334.1 15 - 16 1899 89
NM_181873.3 NP_870988.2 14 - 15 1641 89
XM_006711137.1 XP_006711200.1 14 - 15 1618 89
GenBank mRNA AK097000.1 - 14 - 15 1618 89
AK298775.1 - 15 - 16 1868 89
BC032655.1 - 8 - 9 973 89
BC064372.1 - 13 - 14 1860 89
BC100818.2 - 14 - 15 1559 89
BC100819.2 - 14 - 15 1559 89
BC100820.1 - 14 - 15 1559 89
BC100821.2 - 14 - 15 1512 89

View Full Product Details