Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
Chromosome Location
Chr.1: 149928651 - 149936898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145862.1 NP_001139334.1 9 - 10 1109 57
NM_181873.3 NP_870988.2 8 - 9 851 57
XM_006711137.1 XP_006711200.1 8 - 9 828 57
XM_011509099.2 XP_011507401.1 9 - 10 1002 57
GenBank mRNA AK097000.1 - 8 - 9 828 57
AK296094.1 - 8 - 9 948 57
AK298775.1 - 9 - 10 1078 57
AK304691.1 - 8 - 9 925 57
BC064372.1 - 8 - 9 1237 57
BC100818.2 - 8 - 9 769 57
BC100819.2 - 8 - 9 769 57
BC100820.1 - 8 - 9 769 57
BC100821.2 - 8 - 9 769 57

View Full Product Details