Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZNF33A
- Entrez Gene ID
-
7581
- Gene Name
- zinc finger protein 33A
- Gene Aliases
-
KOX2, KOX31, KOX5, NF11A, ZNF11, ZNF11A, ZNF33, ZZAPK
- Chromosome Location
-
Chr.10: 38010483 - 38067834 on Build GRCh38
View Full Product Details