The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199942.1 | NP_001186871.1 | 30 - 31 | 6360 | 110 |
NM_015018.3 | NP_055833.2 | 30 - 31 | 6387 | 110 | |
XM_011535619.2 | XP_011533921.1 | 29 - 30 | 6352 | 110 | |
XM_017010559.1 | XP_016866048.1 | 30 - 31 | 6382 | 110 | |
XM_017010560.1 | XP_016866049.1 | 29 - 30 | 6310 | 110 | |
XM_017010561.1 | XP_016866050.1 | 30 - 31 | 6334 | 110 | |
XM_017010562.1 | XP_016866051.1 | 29 - 30 | 6283 | 110 | |
XM_017010563.1 | XP_016866052.1 | 30 - 31 | 6361 | 110 | |
XM_017010564.1 | XP_016866053.1 | 30 - 31 | 6361 | 110 | |
XM_017010565.1 | XP_016866054.1 | 30 - 31 | 6355 | 110 | |
XM_017010566.1 | XP_016866055.1 | 30 - 31 | 6355 | 110 | |
XM_017010567.1 | XP_016866056.1 | 29 - 30 | 6283 | 110 | |
XM_017010568.1 | XP_016866057.1 | 29 - 30 | 6352 | 110 | |
XM_017010569.1 | XP_016866058.1 | 28 - 29 | 6269 | 110 | |
XM_017010570.1 | XP_016866059.1 | 29 - 30 | 6283 | 110 | |
XM_017010571.1 | XP_016866060.1 | 29 - 30 | 6277 | 110 | |
XM_017010572.1 | XP_016866061.1 | 30 - 31 | 6382 | 110 | |
XM_017010573.1 | XP_016866062.1 | 30 - 31 | 6382 | 110 | |
XM_017010574.1 | XP_016866063.1 | 30 - 31 | 6382 | 110 | |
GenBank mRNA | AB029040.2 | - | 13 - 14 | 3408 | 110 |
AK027030.1 | - | 10 - 11 | 2154 | 110 | |
AL050080.1 | - | 2 - 3 | 116 | 110 | |
BC048342.1 | - | 4 - 5 | 400 | 110 | |
BC144544.1 | - | 30 - 31 | 6258 | 110 | |
BC150619.1 | - | 30 - 31 | 6258 | 110 | |
BX648667.1 | - | 30 - 31 | 6385 | 110 |