Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42752530-42752789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
700551 c.2639A>T p.K880M Chr.15: 42752607-42752607 on Build GRCh38
700552 c.2683G>C p.D895H Chr.15: 42752563-42752563 on Build GRCh38
5909475 c.2681G>A p.G894E Chr.15: 42752565-42752565 on Build GRCh38
138960 c.2636C>T p.S879F Chr.15: 42752610-42752610 on Build GRCh38
324040 c.2519A>G p.N840S Chr.15: 42752727-42752727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202004988 Chr.15: 42752721-42752721 on Build GRCh38
rs920298246 Chr.15: 42752601-42752601 on Build GRCh38
rs535040519 Chr.15: 42752560-42752560 on Build GRCh38
rs56307230 Chr.15: 42752654-42752654 on Build GRCh38
rs759739645 Chr.15: 42752642-42752642 on Build GRCh38
rs760123245 Chr.15: 42752740-42752740 on Build GRCh38
rs752741012 Chr.15: 42752753-42752753 on Build GRCh38
rs756302462 Chr.15: 42752662-42752662 on Build GRCh38
rs767024763 Chr.15: 42752615-42752615 on Build GRCh38
rs745618805 Chr.15: 42752559-42752559 on Build GRCh38
rs917332444 Chr.15: 42752746-42752746 on Build GRCh38
rs764259278 Chr.15: 42752760-42752760 on Build GRCh38
rs565246063 Chr.15: 42752574-42752574 on Build GRCh38
rs760767810 Chr.15: 42752757-42752757 on Build GRCh38
rs1012848508 Chr.15: 42752611-42752611 on Build GRCh38
rs752082230 Chr.15: 42752638-42752638 on Build GRCh38
rs771925756 Chr.15: 42752573-42752573 on Build GRCh38
rs775057882 Chr.15: 42752736-42752736 on Build GRCh38
rs951685106 Chr.15: 42752621-42752621 on Build GRCh38
rs374795359 Chr.15: 42752558-42752558 on Build GRCh38
rs750216427 Chr.15: 42752666-42752666 on Build GRCh38
rs763332709 Chr.15: 42752614-42752614 on Build GRCh38
rs369689860 Chr.15: 42752711-42752711 on Build GRCh38
rs948795784 Chr.15: 42752751-42752751 on Build GRCh38
rs933276422 Chr.15: 42752561-42752561 on Build GRCh38
rs373859242 Chr.15: 42752671-42752671 on Build GRCh38
rs768128776 Chr.15: 42752579-42752579 on Build GRCh38
rs768078021 Chr.15: 42752744-42752744 on Build GRCh38
rs769356771 Chr.15: 42752723-42752723 on Build GRCh38
rs562383840 Chr.15: 42752639-42752639 on Build GRCh38
rs776320989 Chr.15: 42752589-42752589 on Build GRCh38
rs747638637 Chr.15: 42752722-42752722 on Build GRCh38
rs369978055 Chr.15: 42752613-42752613 on Build GRCh38
rs1035544096 Chr.15: 42752697-42752697 on Build GRCh38
rs567793736 Chr.15: 42752762-42752762 on Build GRCh38
rs984402326 Chr.15: 42752634-42752634 on Build GRCh38
rs776194282 Chr.15: 42752597-42752598 on Build GRCh38
rs746933602 Chr.15: 42752715-42752715 on Build GRCh38
rs377202158 Chr.15: 42752664-42752664 on Build GRCh38
rs761370127 Chr.15: 42752591-42752591 on Build GRCh38
rs768218791 Chr.15: 42752717-42752717 on Build GRCh38
rs769425312 Chr.15: 42752603-42752603 on Build GRCh38
rs867980077 Chr.15: 42752622-42752622 on Build GRCh38
rs771401532 Chr.15: 42752734-42752734 on Build GRCh38
rs746558004 Chr.15: 42752577-42752577 on Build GRCh38
rs908777505 Chr.15: 42752635-42752635 on Build GRCh38
rs772880568 Chr.15: 42752724-42752724 on Build GRCh38
rs767693364 Chr.15: 42752648-42752648 on Build GRCh38
rs773466093 Chr.15: 42752604-42752604 on Build GRCh38
rs757141457 Chr.15: 42752556-42752556 on Build GRCh38
rs758154356 Chr.15: 42752705-42752705 on Build GRCh38
rs763268802 Chr.15: 42752732-42752732 on Build GRCh38

View Full Product Details