Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16197912-16198161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
UniGene
Hs.71040
Gene Chromosome Location
Chr.19: 16197854-16235345 on Build GRCh38
Transcripts
NM_001130524.1 NM_032493.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6294737 c.33G>T p.L11L Chr.19: 16198059-16198059 on Build GRCh38
6294736 c.33G>T p.L11L Chr.19: 16198059-16198059 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763234401 Chr.19: 16198009-16198030 on Build GRCh38
rs185553755 Chr.19: 16197974-16197974 on Build GRCh38
rs757385741 Chr.19: 16198083-16198083 on Build GRCh38
rs769487228 Chr.19: 16198122-16198122 on Build GRCh38
rs766290016 Chr.19: 16197981-16197981 on Build GRCh38
rs758111620 Chr.19: 16198102-16198102 on Build GRCh38
rs190533719 Chr.19: 16197991-16197991 on Build GRCh38
rs746570141 Chr.19: 16198107-16198107 on Build GRCh38
rs745749701 Chr.19: 16198120-16198120 on Build GRCh38
rs946653451 Chr.19: 16198114-16198114 on Build GRCh38
rs778452872 Chr.19: 16198014-16198014 on Build GRCh38
rs746516632 Chr.19: 16198037-16198037 on Build GRCh38
rs900286827 Chr.19: 16197937-16197937 on Build GRCh38
rs774632420 Chr.19: 16198082-16198082 on Build GRCh38
rs1005092035 Chr.19: 16197946-16197946 on Build GRCh38
rs772766260 Chr.19: 16198020-16198020 on Build GRCh38
rs28372758 Chr.19: 16198008-16198009 on Build GRCh38
rs776608508 Chr.19: 16198043-16198043 on Build GRCh38
rs764465021 Chr.19: 16198009-16198010 on Build GRCh38
rs1014872652 Chr.19: 16198024-16198024 on Build GRCh38
rs564852170 Chr.19: 16197997-16197997 on Build GRCh38
rs753640801 Chr.19: 16197982-16197982 on Build GRCh38
rs373507031 Chr.19: 16197943-16197943 on Build GRCh38
rs781722603 Chr.19: 16198009-16198009 on Build GRCh38
rs746262018 Chr.19: 16198010-16198010 on Build GRCh38
rs751757844 Chr.19: 16198080-16198080 on Build GRCh38
rs371239934 Chr.19: 16198127-16198127 on Build GRCh38
rs145442379 Chr.19: 16197975-16197976 on Build GRCh38
rs777929833 Chr.19: 16198001-16198001 on Build GRCh38
rs770946624 Chr.19: 16197995-16197996 on Build GRCh38
rs759342576 Chr.19: 16197985-16197985 on Build GRCh38
rs1050892035 Chr.19: 16197940-16197940 on Build GRCh38
rs944547113 Chr.19: 16198133-16198133 on Build GRCh38
rs746915074 Chr.19: 16197983-16197984 on Build GRCh38
rs151262762 Chr.19: 16198047-16198047 on Build GRCh38
rs765423378 Chr.19: 16197989-16197989 on Build GRCh38
rs752129181 Chr.19: 16198003-16198003 on Build GRCh38
rs763207545 Chr.19: 16198074-16198074 on Build GRCh38
rs756479903 Chr.19: 16198011-16198011 on Build GRCh38
rs952055620 Chr.19: 16198108-16198108 on Build GRCh38
rs181756768 Chr.19: 16198079-16198079 on Build GRCh38
rs776817269 Chr.19: 16198006-16198007 on Build GRCh38
rs770895102 Chr.19: 16198038-16198038 on Build GRCh38
rs775576994 Chr.19: 16198057-16198057 on Build GRCh38
rs770124303 Chr.19: 16198007-16198008 on Build GRCh38
rs143761452 Chr.19: 16198106-16198106 on Build GRCh38
rs886865087 Chr.19: 16198132-16198132 on Build GRCh38
rs762345944 Chr.19: 16198015-16198016 on Build GRCh38
rs562430538 Chr.19: 16197967-16197967 on Build GRCh38
rs770422178 Chr.19: 16198112-16198112 on Build GRCh38
rs771576013 Chr.19: 16198017-16198017 on Build GRCh38
rs201384618 Chr.19: 16198008-16198008 on Build GRCh38
rs747761115 Chr.19: 16198015-16198015 on Build GRCh38
rs11557501 Chr.19: 16198021-16198021 on Build GRCh38
rs765038374 Chr.19: 16198051-16198051 on Build GRCh38
rs1051286508 Chr.19: 16198130-16198130 on Build GRCh38
rs747814120 Chr.19: 16198100-16198100 on Build GRCh38
rs756466184 Chr.19: 16198093-16198093 on Build GRCh38
rs1015553207 Chr.19: 16197949-16197949 on Build GRCh38
rs750205420 Chr.19: 16198119-16198119 on Build GRCh38
rs546375135 Chr.19: 16198126-16198126 on Build GRCh38

View Full Product Details