Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46033308-46033559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7100784 c.2667-1G>T p.? Chr.17: 46033461-46033461 on Build GRCh38
7170419 c.2710G>A p.E904K Chr.17: 46033417-46033417 on Build GRCh38
109857 c.2686C>T p.Q896* Chr.17: 46033441-46033441 on Build GRCh38
1212129 c.2722G>A p.E908K Chr.17: 46033405-46033405 on Build GRCh38
6081121 c.2702G>T p.S901I Chr.17: 46033425-46033425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555819683 Chr.17: 46033444-46033444 on Build GRCh38
rs758257429 Chr.17: 46033374-46033376 on Build GRCh38
rs760027567 Chr.17: 46033486-46033486 on Build GRCh38
rs777976719 Chr.17: 46033391-46033391 on Build GRCh38
rs544544589 Chr.17: 46033380-46033380 on Build GRCh38
rs762584859 Chr.17: 46033427-46033427 on Build GRCh38
rs747126908 Chr.17: 46033392-46033392 on Build GRCh38
rs77009866 Chr.17: 46033522-46033522 on Build GRCh38
rs74867664 Chr.17: 46033429-46033429 on Build GRCh38
rs758751267 Chr.17: 46033385-46033385 on Build GRCh38
rs139615350 Chr.17: 46033409-46033409 on Build GRCh38
rs551968687 Chr.17: 46033412-46033414 on Build GRCh38
rs143533965 Chr.17: 46033441-46033441 on Build GRCh38
rs766948021 Chr.17: 46033463-46033463 on Build GRCh38
rs940272034 Chr.17: 46033519-46033519 on Build GRCh38
rs779148425 Chr.17: 46033373-46033373 on Build GRCh38
rs756578940 Chr.17: 46033366-46033366 on Build GRCh38
rs769568583 Chr.17: 46033419-46033419 on Build GRCh38
rs770946918 Chr.17: 46033397-46033397 on Build GRCh38
rs746717913 Chr.17: 46033475-46033476 on Build GRCh38
rs751132067 Chr.17: 46033364-46033364 on Build GRCh38
rs775332855 Chr.17: 46033424-46033424 on Build GRCh38
rs898679426 Chr.17: 46033538-46033538 on Build GRCh38
rs750568205 Chr.17: 46033352-46033353 on Build GRCh38
rs763594781 Chr.17: 46033352-46033352 on Build GRCh38
rs34859706 Chr.17: 46033536-46033537 on Build GRCh38
rs369804494 Chr.17: 46033371-46033371 on Build GRCh38
rs754301176 Chr.17: 46033469-46033469 on Build GRCh38
rs977269109 Chr.17: 46033367-46033367 on Build GRCh38
rs202001253 Chr.17: 46033395-46033395 on Build GRCh38
rs745808485 Chr.17: 46033412-46033412 on Build GRCh38
rs765619670 Chr.17: 46033491-46033491 on Build GRCh38
rs766865759 Chr.17: 46033370-46033370 on Build GRCh38
rs780281816 Chr.17: 46033518-46033518 on Build GRCh38
rs774020325 Chr.17: 46033440-46033440 on Build GRCh38

View Full Product Details