Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20648438-20648703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PINK1
Entrez Gene ID
65018
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
UniGene
Hs.389171
Gene Chromosome Location
Chr.1: 20633455-20651511 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Gene Symbol
PINK1-AS
Entrez Gene ID
100861548
Gene Name
PINK1 antisense RNA
Gene Aliases
NAPINK1, PINK1-AS1, PINK1AS
UniGene
Hs.710259
Gene Chromosome Location
Chr.1: 20642657-20652193 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3369524 c.1145T>C p.I382T Chr.1: 20648526-20648526 on Build GRCh38
5845326 c.1216G>A p.D406N Chr.1: 20648597-20648597 on Build GRCh38
6312510 c.1243G>T p.A415S Chr.1: 20648624-20648624 on Build GRCh38
5423509 c.1169C>T p.A390V Chr.1: 20648550-20648550 on Build GRCh38
678819 c.1225G>C p.G409R Chr.1: 20648606-20648606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368216507 Chr.1: 20648567-20648567 on Build GRCh38
rs763416852 Chr.1: 20648534-20648534 on Build GRCh38
rs780474701 Chr.1: 20648555-20648556 on Build GRCh38
rs770792215 Chr.1: 20648478-20648478 on Build GRCh38
rs1011308169 Chr.1: 20648515-20648515 on Build GRCh38
rs759427064 Chr.1: 20648483-20648483 on Build GRCh38
rs890075608 Chr.1: 20648507-20648507 on Build GRCh38
rs200462866 Chr.1: 20648540-20648540 on Build GRCh38
rs780102678 Chr.1: 20648581-20648581 on Build GRCh38
rs368939214 Chr.1: 20648559-20648559 on Build GRCh38
rs747239996 Chr.1: 20648593-20648593 on Build GRCh38
rs377471641 Chr.1: 20648682-20648682 on Build GRCh38
rs373068586 Chr.1: 20648654-20648654 on Build GRCh38
rs34203620 Chr.1: 20648511-20648511 on Build GRCh38
rs781434836 Chr.1: 20648659-20648659 on Build GRCh38
rs55962214 Chr.1: 20648521-20648521 on Build GRCh38
rs369152168 Chr.1: 20648469-20648469 on Build GRCh38
rs71653623 Chr.1: 20648675-20648675 on Build GRCh38
rs756873744 Chr.1: 20648637-20648637 on Build GRCh38
rs192131551 Chr.1: 20648642-20648642 on Build GRCh38
rs777439010 Chr.1: 20648557-20648557 on Build GRCh38
rs763185637 Chr.1: 20648605-20648605 on Build GRCh38
rs745739450 Chr.1: 20648485-20648485 on Build GRCh38
rs573411900 Chr.1: 20648619-20648619 on Build GRCh38
rs777060019 Chr.1: 20648526-20648526 on Build GRCh38
rs779917156 Chr.1: 20648652-20648652 on Build GRCh38
rs531300037 Chr.1: 20648506-20648506 on Build GRCh38
rs1050424583 Chr.1: 20648495-20648495 on Build GRCh38
rs959857439 Chr.1: 20648597-20648597 on Build GRCh38
rs202130946 Chr.1: 20648618-20648618 on Build GRCh38
rs374657174 Chr.1: 20648617-20648617 on Build GRCh38
rs371942025 Chr.1: 20648459-20648459 on Build GRCh38
rs45515602 Chr.1: 20648528-20648528 on Build GRCh38
rs760015026 Chr.1: 20648676-20648676 on Build GRCh38
rs370380947 Chr.1: 20648661-20648661 on Build GRCh38
rs758065542 Chr.1: 20648651-20648651 on Build GRCh38
rs534505756 Chr.1: 20648548-20648548 on Build GRCh38
rs772150920 Chr.1: 20648490-20648490 on Build GRCh38
rs119451946 Chr.1: 20648577-20648577 on Build GRCh38
rs372556867 Chr.1: 20648479-20648479 on Build GRCh38
rs548701521 Chr.1: 20648527-20648527 on Build GRCh38
rs139094526 Chr.1: 20648570-20648570 on Build GRCh38
rs145876364 Chr.1: 20648600-20648600 on Build GRCh38
rs758806768 Chr.1: 20648466-20648466 on Build GRCh38
rs763812730 Chr.1: 20648679-20648679 on Build GRCh38
rs369834644 Chr.1: 20648641-20648641 on Build GRCh38
rs377178932 Chr.1: 20648655-20648655 on Build GRCh38
rs747512076 Chr.1: 20648678-20648679 on Build GRCh38
rs774946874 Chr.1: 20648611-20648611 on Build GRCh38
rs574890623 Chr.1: 20648606-20648606 on Build GRCh38
rs145821082 Chr.1: 20648594-20648594 on Build GRCh38
rs757253324 Chr.1: 20648561-20648561 on Build GRCh38
rs45478900 Chr.1: 20648612-20648612 on Build GRCh38
rs752424503 Chr.1: 20648545-20648545 on Build GRCh38
rs556540177 Chr.1: 20648601-20648601 on Build GRCh38
rs373528661 Chr.1: 20648563-20648563 on Build GRCh38
rs45499398 Chr.1: 20648554-20648554 on Build GRCh38
rs776679219 Chr.1: 20648596-20648596 on Build GRCh38
rs532703792 Chr.1: 20648660-20648660 on Build GRCh38
rs200728364 Chr.1: 20648586-20648586 on Build GRCh38
rs376176518 Chr.1: 20648462-20648462 on Build GRCh38
rs768758677 Chr.1: 20648525-20648525 on Build GRCh38
rs1035071310 Chr.1: 20648565-20648565 on Build GRCh38

View Full Product Details