The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000116.4 | NP_000107.1 | 2 - 3 | 560 | 87 |
NM_001303465.1 | NP_001290394.1 | 2 - 3 | 614 | 87 | |
NM_181311.3 | NP_851828.1 | 2 - 3 | 560 | 87 | |
NM_181312.3 | NP_851829.1 | 2 - 3 | 560 | 87 | |
NM_181313.3 | NP_851830.1 | 2 - 3 | 560 | 87 | |
NR_024048.2 | 2 - 3 | 560 | 87 | ||
XM_006724836.1 | XP_006724899.1 | 2 - 3 | 591 | 87 | |
XM_006724837.1 | XP_006724900.1 | 2 - 3 | 591 | 87 | |
XM_006724839.1 | XP_006724902.1 | 2 - 3 | 591 | 87 | |
XM_006724841.3 | XP_006724904.1 | 2 - 3 | 225 | 87 | |
XM_006724842.3 | XP_006724905.1 | 2 - 3 | 225 | 87 | |
XM_011531189.1 | XP_011529491.1 | 2 - 3 | 591 | 87 | |
XM_017029761.1 | XP_016885250.1 | 2 - 3 | 537 | 87 | |
XM_017029762.1 | XP_016885251.1 | 2 - 3 | 591 | 87 | |
XM_017029763.1 | XP_016885252.1 | 2 - 3 | 537 | 87 | |
XM_017029765.1 | XP_016885254.1 | 2 - 3 | 672 | 87 | |
GenBank mRNA | AL531401.3 | - | 2 - 3 | 562 | 87 |
AY231461.1 | - | 2 - 3 | 242 | 87 | |
AY231462.1 | - | 2 - 3 | 242 | 87 | |
AY231463.1 | - | 2 - 3 | 242 | 87 | |
AY231464.1 | - | 2 - 3 | 242 | 87 | |
AY258036.1 | - | 2 - 3 | 242 | 87 | |
AY258037.1 | - | 2 - 3 | 242 | 87 | |
AY258038.1 | - | 2 - 3 | 242 | 87 | |
AY258039.1 | - | 2 - 3 | 242 | 87 | |
BC005062.1 | - | 2 - 3 | 517 | 87 | |
BI822925.1 | - | - | 554 | 87 | |
BX401320.2 | - | 2 - 3 | 400 | 87 | |
CA488175.1 | - | 1 - 2 | 50 | 87 | |
DQ884408.1 | - | 2 - 3 | 288 | 87 | |
KU178349.1 | - | 2 - 3 | 234 | 87 | |
KU178350.1 | - | 2 - 3 | 234 | 87 | |
KU178351.1 | - | 2 - 3 | 234 | 87 | |
KU178352.1 | - | 2 - 3 | 234 | 87 | |
KU178353.1 | - | 2 - 3 | 234 | 87 | |
X92762.1 | - | 2 - 3 | 522 | 87 |