Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16203410-16203681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
UniGene
Hs.71040
Gene Chromosome Location
Chr.19: 16197854-16235345 on Build GRCh38
Transcripts
NM_001130524.1 NM_032493.3 XM_017027420.1 XM_017027421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7269809 c.157G>A p.G53R Chr.19: 16203573-16203573 on Build GRCh38
5513488 c.140C>T p.S47L Chr.19: 16203556-16203556 on Build GRCh38
1391251 c.157delG p.V55fs*182 Chr.19: 16203573-16203573 on Build GRCh38
73688 c.73G>A p.V25M Chr.19: 16203489-16203489 on Build GRCh38
4838973 c.70G>A p.D24N Chr.19: 16203486-16203486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141457321 Chr.19: 16203575-16203575 on Build GRCh38
rs377502937 Chr.19: 16203471-16203471 on Build GRCh38
rs534027489 Chr.19: 16203481-16203481 on Build GRCh38
rs765915345 Chr.19: 16203532-16203534 on Build GRCh38
rs970350647 Chr.19: 16203480-16203480 on Build GRCh38
rs764873016 Chr.19: 16203637-16203638 on Build GRCh38
rs1000353558 Chr.19: 16203628-16203628 on Build GRCh38
rs900683307 Chr.19: 16203576-16203576 on Build GRCh38
rs201225112 Chr.19: 16203601-16203601 on Build GRCh38
rs766789996 Chr.19: 16203469-16203469 on Build GRCh38
rs996411399 Chr.19: 16203650-16203650 on Build GRCh38
rs753324780 Chr.19: 16203572-16203573 on Build GRCh38
rs371850531 Chr.19: 16203585-16203585 on Build GRCh38
rs934999112 Chr.19: 16203533-16203533 on Build GRCh38
rs751224121 Chr.19: 16203615-16203615 on Build GRCh38
rs747628250 Chr.19: 16203452-16203452 on Build GRCh38
rs749993018 Chr.19: 16203641-16203641 on Build GRCh38
rs551396757 Chr.19: 16203439-16203439 on Build GRCh38
rs781205990 Chr.19: 16203579-16203579 on Build GRCh38
rs944338208 Chr.19: 16203574-16203574 on Build GRCh38
rs369281605 Chr.19: 16203658-16203658 on Build GRCh38
rs780111532 Chr.19: 16203540-16203540 on Build GRCh38
rs780166635 Chr.19: 16203652-16203652 on Build GRCh38
rs571457066 Chr.19: 16203453-16203453 on Build GRCh38
rs374334160 Chr.19: 16203590-16203590 on Build GRCh38
rs765514475 Chr.19: 16203609-16203609 on Build GRCh38
rs762029617 Chr.19: 16203573-16203573 on Build GRCh38
rs749149339 Chr.19: 16203556-16203556 on Build GRCh38
rs756989673 Chr.19: 16203633-16203633 on Build GRCh38
rs746086930 Chr.19: 16203594-16203594 on Build GRCh38
rs756722435 Chr.19: 16203487-16203487 on Build GRCh38
rs1050012109 Chr.19: 16203600-16203600 on Build GRCh38
rs117647142 Chr.19: 16203488-16203488 on Build GRCh38
rs769839602 Chr.19: 16203499-16203499 on Build GRCh38
rs554289228 Chr.19: 16203485-16203485 on Build GRCh38
rs1024279051 Chr.19: 16203449-16203449 on Build GRCh38
rs200460904 Chr.19: 16203486-16203486 on Build GRCh38
rs200294719 Chr.19: 16203606-16203606 on Build GRCh38
rs927596999 Chr.19: 16203523-16203523 on Build GRCh38
rs146546344 Chr.19: 16203572-16203572 on Build GRCh38
rs755561939 Chr.19: 16203644-16203644 on Build GRCh38
rs904686319 Chr.19: 16203618-16203618 on Build GRCh38
rs146062531 Chr.19: 16203557-16203557 on Build GRCh38
rs766957694 Chr.19: 16203578-16203578 on Build GRCh38
rs372560384 Chr.19: 16203489-16203489 on Build GRCh38
rs112581784 Chr.19: 16203635-16203635 on Build GRCh38
rs754685058 Chr.19: 16203573-16203573 on Build GRCh38
rs969307658 Chr.19: 16203472-16203472 on Build GRCh38

View Full Product Details