Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16986869-16987110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6059553 c.2902G>T p.V968L Chr.1: 16987095-16987095 on Build GRCh38
5989559 c.3046C>A p.Q1016K Chr.1: 16987083-16987083 on Build GRCh38
6888421 c.3092C>T p.P1031L Chr.1: 16986948-16986948 on Build GRCh38
1295497 c.3138G>C p.E1046D Chr.1: 16986902-16986902 on Build GRCh38
3478449 c.3087C>T p.F1029F Chr.1: 16986953-16986953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749123699 Chr.1: 16986922-16986922 on Build GRCh38
rs768922216 Chr.1: 16986963-16986963 on Build GRCh38
rs184878897 Chr.1: 16986953-16986953 on Build GRCh38
rs767135194 Chr.1: 16986979-16986979 on Build GRCh38
rs779969316 Chr.1: 16987090-16987090 on Build GRCh38
rs745888153 Chr.1: 16986936-16986936 on Build GRCh38
rs867009100 Chr.1: 16987088-16987088 on Build GRCh38
rs531265631 Chr.1: 16987032-16987032 on Build GRCh38
rs763544862 Chr.1: 16987040-16987040 on Build GRCh38
rs748408338 Chr.1: 16987031-16987031 on Build GRCh38
rs759878626 Chr.1: 16987025-16987028 on Build GRCh38
rs755900033 Chr.1: 16986911-16986911 on Build GRCh38
rs538236343 Chr.1: 16986929-16986929 on Build GRCh38
rs56165294 Chr.1: 16987013-16987013 on Build GRCh38
rs367760588 Chr.1: 16987068-16987068 on Build GRCh38
rs768797555 Chr.1: 16987019-16987019 on Build GRCh38
rs56299881 Chr.1: 16986972-16986972 on Build GRCh38
rs771603791 Chr.1: 16986973-16986973 on Build GRCh38
rs747103119 Chr.1: 16987016-16987016 on Build GRCh38
rs773787370 Chr.1: 16986976-16986976 on Build GRCh38
rs750963832 Chr.1: 16986895-16986895 on Build GRCh38
rs139462252 Chr.1: 16987047-16987047 on Build GRCh38
rs7531163 Chr.1: 16986959-16986959 on Build GRCh38
rs772117418 Chr.1: 16986944-16986944 on Build GRCh38
rs765632065 Chr.1: 16987072-16987072 on Build GRCh38
rs367885671 Chr.1: 16987012-16987012 on Build GRCh38
rs777252902 Chr.1: 16987017-16987018 on Build GRCh38
rs951420437 Chr.1: 16987061-16987061 on Build GRCh38
rs368908107 Chr.1: 16986966-16986966 on Build GRCh38
rs55885047 Chr.1: 16987022-16987022 on Build GRCh38
rs373946665 Chr.1: 16986993-16986993 on Build GRCh38
rs765075611 Chr.1: 16987074-16987074 on Build GRCh38
rs200537435 Chr.1: 16987086-16987086 on Build GRCh38
rs200045802 Chr.1: 16986991-16986991 on Build GRCh38
rs56126202 Chr.1: 16986896-16986896 on Build GRCh38
rs773521014 Chr.1: 16987036-16987036 on Build GRCh38
rs906992159 Chr.1: 16986960-16986960 on Build GRCh38
rs188473562 Chr.1: 16986986-16986986 on Build GRCh38
rs757076945 Chr.1: 16986928-16986928 on Build GRCh38
rs763606239 Chr.1: 16987055-16987055 on Build GRCh38
rs958645946 Chr.1: 16987020-16987020 on Build GRCh38
rs777700606 Chr.1: 16986914-16986914 on Build GRCh38
rs892885782 Chr.1: 16986932-16986932 on Build GRCh38
rs765618139 Chr.1: 16986967-16986967 on Build GRCh38
rs771581490 Chr.1: 16987044-16987044 on Build GRCh38
rs371344553 Chr.1: 16986987-16986987 on Build GRCh38
rs202166353 Chr.1: 16987089-16987089 on Build GRCh38
rs374690625 Chr.1: 16987048-16987048 on Build GRCh38
rs752317109 Chr.1: 16986981-16986981 on Build GRCh38
rs372182128 Chr.1: 16987070-16987070 on Build GRCh38
rs766447629 Chr.1: 16986905-16986905 on Build GRCh38
rs757098756 Chr.1: 16986990-16986990 on Build GRCh38
rs146884019 Chr.1: 16986909-16986909 on Build GRCh38
rs368292341 Chr.1: 16986977-16986977 on Build GRCh38

View Full Product Details