Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR3
Entrez Gene ID
10096
Gene Name
ARP3 actin related protein 3 homolog
Gene Aliases
ARP3
Chromosome Location
Chr.2: 113889934 - 113961554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277140.1 NP_001264069.1 6 - 7 678 92
NM_005721.4 NP_005712.1 6 - 7 887 92
NR_102318.1 5 - 6 762 92
GenBank mRNA AB209353.1 - 4 - 5 396 92
AF006083.1 - 6 - 7 713 92
AF127773.1 - 6 - 7 756 92
AK300030.1 - 5 - 6 586 92
AK300224.1 - 5 - 6 508 92
AK302152.1 - 6 - 7 632 92
AK302359.1 - 6 - 7 666 92
AK312659.1 - 6 - 7 687 92
AK316213.1 - 6 - 7 709 92
AK316396.1 - 6 - 7 678 92
BC044590.1 - 6 - 7 861 92

View Full Product Details