Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP12-2
Entrez Gene ID
353323
Gene Name
keratin associated protein 12-2
Gene Aliases
KAP12.2, KRTAP12.2
Chromosome Location
Chr.21: 44666189 - 44666927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181684.2 NP_859012.1 1 - 1 152 147
GenBank mRNA AB076362.1 - 1 - 1 131 147
AJ566389.1 - 1 - 1 161 147
BC120941.1 - 1 - 1 140 147

View Full Product Details