Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LONRF2
Entrez Gene ID
164832
Gene Name
LON peptidase N-terminal domain and ring finger 2
Gene Aliases
RNF192
Chromosome Location
Chr.2: 100273291 - 100322733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198461.3 NP_940863.3 6 - 7 1996 68
GenBank mRNA AK127206.1 - 6 - 7 698 68
BC101662.1 - 5 - 6 655 68
BC101664.1 - 5 - 6 655 68
BC143468.1 - 5 - 6 655 68
DQ020495.1 - 6 - 7 1996 68

View Full Product Details