The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 73 - 74 | 11884 | 104 |
XM_011533466.1 | XP_011531768.1 | 73 - 74 | 11869 | 104 | |
XM_011533467.1 | XP_011531769.1 | 73 - 74 | 11836 | 104 | |
XM_011533468.1 | XP_011531770.1 | 73 - 74 | 11806 | 104 | |
XM_011533469.1 | XP_011531771.1 | 72 - 73 | 11728 | 104 | |
XM_017005861.1 | XP_016861350.1 | 72 - 73 | 12552 | 104 | |
XM_017005862.1 | XP_016861351.1 | 52 - 53 | 8444 | 104 | |
GenBank mRNA | AK097746.1 | - | 13 - 14 | 2032 | 104 |
AK128592.1 | - | 21 - 22 | 3310 | 104 |