The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135052.3 | NP_001128524.1 | 8 - 9 | 1370 | 144 |
NM_001174167.2 | NP_001167638.1 | 9 - 10 | 1385 | 144 | |
NM_001174168.2 | NP_001167639.1 | 8 - 9 | 1235 | 144 | |
NM_003177.6 | NP_003168.2 | 9 - 10 | 1439 | 144 | |
XM_005252147.3 | XP_005252204.1 | 9 - 10 | 1350 | 144 | |
XM_011518946.2 | XP_011517248.1 | 9 - 10 | 1299 | 144 | |
GenBank mRNA | AK075020.1 | - | 5 - 6 | 503 | 144 |
AK290927.1 | - | 9 - 10 | 1283 | 144 | |
BC001645.2 | - | 9 - 10 | 1331 | 144 | |
BC002962.1 | - | 9 - 10 | 1331 | 144 | |
BC011399.1 | - | 8 - 9 | 1219 | 144 | |
BX647192.1 | - | 2 - 3 | 2459 | 144 | |
FN391891.1 | - | 8 - 9 | 1217 | 144 | |
FN391892.1 | - | 8 - 9 | 1221 | 144 | |
FN391893.1 | - | 8 - 9 | 1180 | 144 | |
L28824.1 | - | 9 - 10 | 1331 | 144 | |
S80267.1 | - | 8 - 9 | 1185 | 144 | |
X73568.1 | - | 9 - 10 | 1323 | 144 | |
Z29630.1 | - | 8 - 9 | 1263 | 144 |