Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC17
Entrez Gene ID
149483
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
Chromosome Location
Chr.1: 45620044 - 45624787 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114938.2 NP_001108410.2 9 - 10 1336 78
NM_001190182.1 NP_001177111.1 9 - 10 1309 78
XM_011540812.1 XP_011539114.1 8 - 9 2163 78
XM_011540814.1 XP_011539116.1 8 - 9 2163 78
XM_017000450.1 XP_016855939.1 8 - 9 2136 78
XM_017000451.1 XP_016855940.1 7 - 8 2067 78
XM_017000452.1 XP_016855941.1 9 - 10 2064 78
XM_017000453.1 XP_016855942.1 7 - 8 2055 78
XM_017000454.1 XP_016855943.1 8 - 9 1234 78
XM_017000455.1 XP_016855944.1 7 - 8 781 78
XM_017000456.1 XP_016855945.1 7 - 8 782 78
XM_017000458.1 XP_016855947.1 7 - 8 2044 78
GenBank mRNA AK057646.1 - 8 - 9 1239 78
AK298104.1 - 9 - 10 1307 78
AK303939.1 - 9 - 10 1334 78
BC029888.1 - 7 - 8 959 78
BC128177.1 - 7 - 8 925 78

View Full Product Details