Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLUHP3
Entrez Gene ID
100132341
Gene Name
clustered mitochondria homolog pseudogene 3
Gene Aliases
C16orf67, KIAA0664L3, KIAA0664P3, URLC3
Chromosome Location
Chr.16: 31700613 - 31707437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024034.1 4 - 5 683 65
GenBank mRNA AB101205.1 - 4 - 5 706 65
AK054698.1 - 2 - 3 361 65
BC000552.1 - 3 - 4 474 65
BC021555.1 - 4 - 5 683 65


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA054329.1 - 2 - 3 361 Chr. 16 - 31700595 - 31705398 [+] on Build GRCh38 65
GenBankmRNA DA591583.1 - 3 - 4 521 Chr. 16 - 31700579 - 31704274 [+] on Build GRCh38 65
GenBankmRNA BP198043.1 - 3 - 4 521 Chr. 16 - 31700579 - 31704303 [+] on Build GRCh38 65
GenBankmRNA CK824943.1 - 2 - 3 323 Chr. 16 - 31700630 - 31704342 [+] on Build GRCh38 65
GenBankmRNA DB119181.1 - 3 - 4 493 Chr. 16 - 31700607 - 31704299 [+] on Build GRCh38 65
GenBankmRNA DA969723.1 - 3 - 4 507 Chr. 16 - 31700595 - 31704274 [+] on Build GRCh38 65
GenBankmRNA BQ053459.1 - 3 - 4 488 Chr. 16 - 31700612 - 31705775 [+] on Build GRCh38 65
GenBankmRNA DA017429.1 - 3 - 4 514 Chr. 16 - 31700581 - 31704266 [+] on Build GRCh38 65
GenBankmRNA DB115556.1 - 3 - 4 512 Chr. 16 - 31700588 - 31704271 [+] on Build GRCh38 65
GenBankmRNA BM272077.1 - 2 - 3 257 Chr. 16 - 31700696 - 31704323 [+] on Build GRCh38 65
GenBankmRNA AL706241.1 - 2 - 3 384 Chr. 16 - 31700570 - 31705745 [+] on Build GRCh38 65
GenBankmRNA BG257678.1 - 3 - 4 468 Chr. 16 - 31700632 - 31705409 [+] on Build GRCh38 65
GenBankmRNA BQ708121.1 - 3 - 4 449 Chr. 16 - 31700651 - 31705939 [+] on Build GRCh38 65
GenBankmRNA DB222656.1 - 3 - 4 506 Chr. 16 - 31700595 - 31704263 [+] on Build GRCh38 65
GenBankmRNA DA071570.1 - 2 - 3 351 Chr. 16 - 31700602 - 31705400 [+] on Build GRCh38 65
GenBankmRNA BP200087.1 - 2 - 3 Chr. 16 - 31700595 - 31704326 [+] on Build GRCh38 64
GenBankmRNA CA489587.1 - 4 - 5 674 Chr. 16 - 31700622 - 31704390 [+] on Build GRCh38 65
GenBankmRNA BE313941.1 - 3 - 4 446 Chr. 16 - 31700655 - 31704300 [+] on Build GRCh38 65
GenBankmRNA BE297853.1 - 3 - 4 509 Chr. 16 - 31700613 - 31704390 [+] on Build GRCh38 65
GenBankmRNA DN992043.1 - 3 - 4 490 Chr. 16 - 31700617 - 31704379 [+] on Build GRCh38 65
GenBankmRNA CV029253.1 - 200 Chr. - - [-] on Build 65
GenBankmRNA BP196478.1 - 3 - 4 487 Chr. 16 - 31700613 - 31704315 [+] on Build GRCh38 65
GenBankmRNA CV029207.1 - 200 Chr. - - [-] on Build 65
GenBankmRNA BE270758.1 - 3 - 4 511 Chr. 16 - 31700616 - 31705386 [+] on Build GRCh38 65
GenBankmRNA DA965407.1 - 3 - 4 529 Chr. 16 - 31700572 - 31704260 [+] on Build GRCh38 65
GenBankmRNA DB073280.1 - 2 - 3 359 Chr. 16 - 31700594 - 31704390 [+] on Build GRCh38 65


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Y-box binding protein 3 pseudogene 1 YBX3P1 CSDAP1 -133.6kb Anti-sense Chr. - 31567767 - 31569524 on Build GRCh38
Downstream zinc finger protein 720 ZNF720 8.8kb Sense Chr. - 31713229 - 31761565 on Build GRCh38

View Full Product Details