The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003280.2 | NP_003271.1 | 2 - 3 | 81 | 66 |
GenBank mRNA | AF020769.1 | - | 2 - 3 | 59 | 66 |
AK313743.1 | - | 2 - 3 | 81 | 66 | |
BC030244.1 | - | 2 - 3 | 68 | 66 | |
CR541865.1 | - | 2 - 3 | 55 | 66 | |
CR541885.1 | - | 2 - 3 | 55 | 66 | |
X07897.1 | - | 2 - 3 | 81 | 66 |