The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099436.3 | NP_001092906.3 | 13 - 14 | 1387 | 88 |
NM_001284364.2 | NP_001271293.2 | 13 - 14 | 1387 | 88 | |
NM_001284365.2 | NP_001271294.1 | 13 - 14 | 1310 | 88 | |
NR_104307.2 | 12 - 13 | 1568 | 88 | ||
NR_104308.2 | 12 - 13 | 1564 | 84 | ||
NR_104309.2 | 12 - 13 | 1549 | 69 | ||
XM_005254289.2 | XP_005254346.1 | 14 - 15 | 1402 | 88 | |
XM_017022068.1 | XP_016877557.1 | 14 - 15 | 1436 | 88 | |
XM_017022069.1 | XP_016877558.1 | 14 - 15 | 1440 | 88 | |
XM_017022070.1 | XP_016877559.1 | 14 - 15 | 1453 | 88 | |
XM_017022071.1 | XP_016877560.1 | 14 - 15 | 1440 | 88 | |
XM_017022072.1 | XP_016877561.1 | 13 - 14 | 1616 | 88 | |
GenBank mRNA | AK293181.1 | - | 13 - 14 | 1240 | 88 |
AK294241.1 | - | 13 - 14 | 1321 | 88 | |
AK294245.1 | - | 13 - 14 | 1317 | 88 | |
AK294251.1 | - | 13 - 14 | 1321 | 88 | |
AK298879.1 | - | 13 - 14 | 1244 | 88 | |
AK299380.1 | - | 13 - 14 | 1314 | 88 | |
AL117482.1 | - | 12 - 13 | 1703 | 88 | |
AL360256.1 | - | 8 - 9 | 980 | 88 | |
BC048289.1 | - | 12 - 13 | 1492 | 84 | |
BC056423.1 | - | 12 - 13 | 1467 | 69 | |
BC157884.1 | - | 13 - 14 | 1369 | 88 |