Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFS1
Entrez Gene ID
9054
Gene Name
NFS1 cysteine desulfurase
Gene Aliases
HUSSY-08, IscS, NIFS
Chromosome Location
Chr.20: 35668688 - 35699365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198989.1 NP_001185918.1 5 - 6 580 83
NM_021100.4 NP_066923.3 6 - 7 733 83
NR_037570.2 7 - 8 855 83
GenBank mRNA AF097025.2 - 6 - 7 719 83
AJ010952.1 - 2 - 3 241 83
AK001265.1 - 6 - 7 719 83
AK001470.1 - 6 - 7 729 83
AK056242.1 - 6 - 7 1004 83
AK223239.1 - 6 - 7 719 83
AK297969.1 - 7 - 8 842 83
AK302023.1 - 5 - 6 567 83
AK312293.1 - 6 - 7 720 83
BC065560.1 - 6 - 7 705 83

View Full Product Details