The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139126.3 | NP_624311.1 | 7 - 8 | 769 | 149 |
GenBank mRNA | AF357880.1 | - | 7 - 8 | 702 | 149 |
AK092069.1 | - | 7 - 8 | 702 | 149 | |
AK315388.1 | - | 7 - 8 | 769 | 149 | |
BC020986.1 | - | 7 - 8 | 705 | 149 | |
BC064134.1 | - | 7 - 8 | 687 | 149 | |
BC107684.1 | - | 7 - 8 | 703 | 149 | |
BX537536.1 | - | 7 - 8 | 704 | 149 |