The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018211.3 | NP_060681.2 | 2 - 3 | 389 | 107 |
XM_006710738.3 | XP_006710801.2 | 2 - 3 | 496 | 107 | |
XM_011541706.2 | XP_011540008.1 | 2 - 3 | 505 | 107 | |
GenBank mRNA | AB046799.1 | - | 2 - 3 | 389 | 107 |